missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Rat IL-2 Protein

Product Code. 18632506 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
This item is not returnable. View return policy

Product Code. 18632506

Brand: Novus Biologicals™ NBP2352325ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

IL2 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine CTLL-2 cells is less than 0.2 ng/mL, corresponding to a specific activity of > 5.0 x 10^6 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P17108, P17108, P17108
For Use With (Application) SDS-PAGE, Bioactivity
Formulation 0.2 μm filtered concentrated sterile solution in 10 mM sodium citrate, pH 4.0.
Gene ID (Entrez) 3558
Molecular Weight (g/mol) MolecularWeight-theroretical: 15.5 kDa
Quantity 5 μg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 1 EU/μg of IL-2 as determined by LAL method.
Gene Alias aldesleukin, IL-2, IL-2 lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Gene Symbol IL2
Research Category Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers
Purity or Quality Grade >97 % pure by SDS-PAGE and HPLC
Protein IL-2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.