missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat IL-2 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
IL2 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine CTLL-2 cells is less than 0.2 ng/mL, corresponding to a specific activity of > 5.0 x 10^6 IU/mg.
Specifications
Specifications
| Accession Number | P17108, P17108, P17108 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | 0.2 μm filtered concentrated sterile solution in 10 mM sodium citrate, pH 4.0. |
| Gene ID (Entrez) | 3558 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 15.5 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of IL-2 as determined by LAL method. |
| Gene Alias | aldesleukin, IL-2, IL-2 lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF |
| Gene Symbol | IL2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction