missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat HRAS / GTPase Hras Protein

A DNA sequence encoding the Rattus norvegicus (Rat) HRAS / GTPase Hras, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8837-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16096741

  • 2990.00€ / 1mg

Estimated Shipment: 05-06-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001091711.1
HRAS / GTPase Hras Protein
1 mg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
293621
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Hras
Recombinant Protein
E. coli
TEYKLVVVGA GGVGKSALTI QLIQNHFVDE YDPTIEDSYR KQVVIDGETC LLDILDTAGQ EEYSAMRDQY MRTGEGFLCV FAINNTKSFE DIHQYREQIK RVKDSDDVPM VLVGNKCDLA ARTVESRQAQ DLARSYGIPY IETSAKTRQG VEDAFYTLVR EIRQHKLRKL NPPDESGPGC MSCKC
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers