missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat HRAS / GTPase Hras Protein
A DNA sequence encoding the Rattus norvegicus (Rat) HRAS / GTPase Hras, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8837-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001091711.1 | |
HRAS / GTPase Hras Protein | |
Research Use Only | |
Hras | |
Recombinant Protein | |
E. coli | |
TEYKLVVVGA GGVGKSALTI QLIQNHFVDE YDPTIEDSYR KQVVIDGETC LLDILDTAGQ EEYSAMRDQY MRTGEGFLCV FAINNTKSFE DIHQYREQIK RVKDSDDVPM VLVGNKCDLA ARTVESRQAQ DLARSYGIPY IETSAKTRQG VEDAFYTLVR EIRQHKLRKL NPPDESGPGC MSCKC | |
Greater than 90% as determined by SDS-PAGE. |
293621 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |