missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat HRAS / GTPase Hras Protein

A DNA sequence encoding the Rattus norvegicus (Rat) HRAS / GTPase Hras, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8837-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026751

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001091711.1
HRAS / GTPase Hras Protein
Research Use Only
Hras
Recombinant Protein
E. coli
TEYKLVVVGA GGVGKSALTI QLIQNHFVDE YDPTIEDSYR KQVVIDGETC LLDILDTAGQ EEYSAMRDQY MRTGEGFLCV FAINNTKSFE DIHQYREQIK RVKDSDDVPM VLVGNKCDLA ARTVESRQAQ DLARSYGIPY IETSAKTRQG VEDAFYTLVR EIRQHKLRKL NPPDESGPGC MSCKC
Greater than 90% as determined by SDS-PAGE.
293621
50 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers