Learn More
enQuireBio™ Recombinant Rat HRAS / GTPase Hras Protein
A DNA sequence encoding the Rattus norvegicus (Rat) HRAS / GTPase Hras, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP8837-ec-500ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16016751
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Specifications
NP_001091711.1
HRAS / GTPase Hras Protein
Research Use Only
Hras
Recombinant Protein
E. coli
TEYKLVVVGA GGVGKSALTI QLIQNHFVDE YDPTIEDSYR KQVVIDGETC LLDILDTAGQ EEYSAMRDQY MRTGEGFLCV FAINNTKSFE DIHQYREQIK RVKDSDDVPM VLVGNKCDLA ARTVESRQAQ DLARSYGIPY IETSAKTRQG VEDAFYTLVR EIRQHKLRKL NPPDESGPGC MSCKC
Greater than 90% as determined by SDS-PAGE.
293621
500 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol