missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat beta-NGF Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Unit Size:
100µg
Description
The Recombinant Rat beta-NGF Protein is derived from Sf 21 (baculovirus). The Recombinant Rat beta-NGF Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and NaCl. |
| Gene ID (Entrez) | 310738 |
| Molecular Weight (g/mol) | 13.2 kDa (monomer) |
| Name | beta-NGF |
| Quantity | 100 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived rat beta-NGF protein Ser122-Gly241 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction