Learn More
enQuireBio™ Recombinant Rat beta-NGF / Beta-NGF Protein
Product Code. p-7155497
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16086951
Brand: enQuireBio™ QP8881ec10ug
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
A DNA sequence encoding the Rattus norvegicus (Rat) beta-NGF / Beta-NGF Protein, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_001263984.1
Gene ID (Entrez) 310738
Name beta-NGF / Beta-NGF Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Ngf
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag His
Sequence THPVFHMGEF SVCDSVSVWV GDKTTATDIK GKEVTVLGEV NINNSVFKQY FFETKCRAPN PVESGCRGID SKHWNSYCTT THTFVKALTT DDKQAAWRFI RIDTACVCVL SRKAARRG
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.