missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Anionic trypsin-1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Anionic trypsin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6536-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064010

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_036767.1
Anionic trypsin-1 Protein
Research Use Only
Prss1
Recombinant Protein
E. coli
IVGGYTCPEH SVPYQVSLNS GYHFCGGSLI NDQWVVSAAH CYKSRIQVRL GEHNINVLEG DEQFINAAKI IKHPNYSSWT LNNDIMLIKL SSPVKLNARV APVALPSACA PAGTQCLISG WGNTLSNGVN NPDLLQCVDA PVLSQADCEA AYPGEITSSM ICVGFLEGGK DSCQGDSGGP VVCNGQLQGI VSWGYGCALP DNPGVYTKVC NFVGWIQDTI AAN
Greater than 90% as determined by SDS-PAGE.
24691
200 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers