missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Anionic trypsin-1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Anionic trypsin-1, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_036767.1 |
---|---|
Gene ID (Entrez) | 24691 |
Name | Anionic trypsin-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16034010
|
enQuireBio™
QP6536-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16054010
|
enQuireBio™
QP6536-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16044010
|
enQuireBio™
QP6536-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16064010
|
enQuireBio™
QP6536-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16074010
|
enQuireBio™
QP6536-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16084010
|
enQuireBio™
QP6536-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_036767.1 | |
Anionic trypsin-1 Protein | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
24691 | |
Research Use Only | |
Prss1 | |
Recombinant Protein | |
E. coli | |
IVGGYTCPEH SVPYQVSLNS GYHFCGGSLI NDQWVVSAAH CYKSRIQVRL GEHNINVLEG DEQFINAAKI IKHPNYSSWT LNNDIMLIKL SSPVKLNARV APVALPSACA PAGTQCLISG WGNTLSNGVN NPDLLQCVDA PVLSQADCEA AYPGEITSSM ICVGFLEGGK DSCQGDSGGP VVCNGQLQGI VSWGYGCALP DNPGVYTKVC NFVGWIQDTI AAN | |
Greater than 90% as determined by SDS-PAGE. |