missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pseudomonas Thiol:disulfide interchange protein DsbA Protein
A DNA sequence encoding the Pseudomonas syringae pv. tomato (strain DC3000) Thiol:disulfide interchange protein DsbA, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_790191.1 |
---|---|
Gene ID (Entrez) | 1181950 |
Name | Pseudomonas Thiol:disulfide interchange protein DsbA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16009010
|
enQuireBio™
QP7497-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16029010
|
enQuireBio™
QP7497-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16019010
|
enQuireBio™
QP7497-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16039010
|
enQuireBio™
QP7497-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16049010
|
enQuireBio™
QP7497-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16059010
|
enQuireBio™
QP7497-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_790191.1 | |
Pseudomonas Thiol:disulfide interchange protein DsbA Protein | |
Not determined. | |
Full Length | |
Pseudomonas | |
His | |
Tris-based buffer, 50% glycerol |
1181950 | |
Research Use Only | |
dsbA | |
Recombinant Protein | |
E. coli | |
AEPIESGKQY VELTSAVPVA VPGKIEVIEL FWYGCPHCYA FEPTINPWVE KLPSDVNFVR IPAMFGGPWD AHGQLFITLD TMGVEHKVHA AVFEAIQKGG KRLTDKNDMA DFVATQGVNK DDFLKTFDSF AVKGKIAQYK ELAKKYEVTG VPTMIVNGKY RFDLGSAGGP EKTLQVADQL IDKERAAAKA AK | |
Greater than 90% as determined by SDS-PAGE. |