missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pseudomonas Thiol:disulfide interchange protein DsbA Protein
A DNA sequence encoding the Pseudomonas syringae pv. tomato (strain DC3000) Thiol:disulfide interchange protein DsbA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7497-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_790191.1 | |
Pseudomonas Thiol:disulfide interchange protein DsbA Protein | |
Research Use Only | |
dsbA | |
Recombinant Protein | |
E. coli | |
AEPIESGKQY VELTSAVPVA VPGKIEVIEL FWYGCPHCYA FEPTINPWVE KLPSDVNFVR IPAMFGGPWD AHGQLFITLD TMGVEHKVHA AVFEAIQKGG KRLTDKNDMA DFVATQGVNK DDFLKTFDSF AVKGKIAQYK ELAKKYEVTG VPTMIVNGKY RFDLGSAGGP EKTLQVADQL IDKERAAAKA AK | |
Greater than 90% as determined by SDS-PAGE. |
1181950 | |
200 μg | |
Not determined. | |
Full Length | |
Pseudomonas | |
His | |
Tris-based buffer, 50% glycerol |