Learn More
enQuireBio™ Recombinant P. gingivalis Gingipain R2 Protein
Product Code. p-7153809
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16056930
Brand: enQuireBio™ QP698910ug
A DNA sequence encoding the Porphyromonas gingivalis (strain ATCC BAA-308 / W83) Gingipain R2, was expressed in the hosts and tags indicated.
Specifications
Accession Number WP_010956050.1
Name P. gingivalis Gingipain R2 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol rgpB
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity P. gingivalis
Species E. coli
Protein Tag His-SUMO
Sequence YTPVEEKENG RMIVIVPKKY EEDIEDFVDW KNQRGLRTEV KVAEDIASPV TANAIQQFVK QEYEKEGNDL TYVLLVGDHK DIPAKITPGI KSDQVYGQIV GNDHYNEVFI GRFSCESKED LKTQIDRTIH YERNITTEDK WLGQALCIAS AEGGPSADNG ESDIQHENII ANLLTQYGYT KIIKCYDPGV TPKNIIDAFN GGISLANYTG HGSETAWGTS HFGTTHVKQL TNSNQLPFIF DVAC
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.