missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant P. aeruginosa Beta-lactamase Protein

Product Code. 16087420
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16087420

Brand: enQuireBio™ QP711910ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Beta-lactamase, was expressed in the hosts and tags indicated. option, ab

Specifications

Accession Number NP_252799.1
Gene ID (Entrez) 878149
Name P. aeruginosa Beta-lactamase Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ampC
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity P. aeruginosa
Species E. coli
Protein Tag His-SUMO
Sequence GEAPADRLKA LVDAAVQPVM KANDIPGLAV AISLKGEPHY FSYGLASKED GRRVTPETLF EIGSVSKTFT ATLAGYALTQ DKMRLDDRAS QHWPALQGSR FDGISLLDLA TYTAGGLPLQ FPDSVQKDQA QIRDYYRQWQ PTYAPGSQRL YSNPSIGLFG YLAARSLGQP FERLMEQQVF PALGLEQTHL DVPEAALAQY AQGYGKDDRP LRVGPGPLDA EGYGVKTSAA DLLRFVDANL HPERLDRPWA QALDATHRGY YKVGDMTQGL GWEAYDWPIS LKRLQAGNST PMALQPHRIA RLPAPQALEG QRLLNKTGST NGFGAYVAFV PGRDLGLVIL ANRNYPNAER VKIAYAILSG LEQQGKVPLK R
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.