missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other PhI p 6 Protein

Product Code. p-7150925
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15992329 50 μg 50µg
15972329 100 μg 100µg
15982329 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15992329 Supplier enQuireBio™ Supplier No. QP1304350ug
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A cDNA sequence encoding the PhI p 6 was constructed and used to recombinantly synthesize the protein.

Specifications

Name PhI p 6 Protein
Quantity 50 μg
Regulatory Status RUO
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
Species Baculovirus
Protein Tag His
Sequence MVAMFLAVAV VLGLATSPTA EGGKATTEEQ KLIEDVNASF RAAMATTANV PPADKYKTFE AAFTVSSKRN LADAVSKAPQ LVPKLDEVYN AAYNAADHAA PEDKYEAFVL HFSEALRIIA GTPEVHAVKP GA
Buffer PhI p 6 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Purity or Quality Grade Greater than 80.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.