missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other HSA Lipid free Protein
Specifications
Specifications
| Gene ID (Entrez) | 213 |
| Name | HSA Lipid free Protein |
| Quantity | 100 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Gene Symbol | ALB |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | Rice |
| Protein Tag | Untagged |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?