missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Zinc finger protein 346 Protein
A DNA sequence encoding the Mus musculus (Mouse) Zinc finger protein 346, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6908-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036147.1 | |
Zinc finger protein 346 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
26919 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Znf346 | |
Recombinant Protein | |
E. coli | |
MECPAPDATD AADPGEAGPY KGSEEPEGRE PDGVRFDRER ARRLWEAVSG AQPAGREEVE HMIQKNQCLF TSTQCKVCCA MLISESQKLA HYQSKKHANK VKRYLAIHGM ETIKGDVKRL DSDQKSSRSK DKNHCCPICN MTFSSPAVAQ SHYLGKTHAK SLKLKQQSTK GAALQQNREM LDPDKFCSLC HSTFNDPAMA QQHYMGKRHR KQETKLKLMA HYGRLADPAV SDLPAGKGYP CKTCKIVLNS IEQYQAHVSG FKHKNQSPKT LVTLGSQTPV QTQPTPKDSS TVQD |