missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Vomeronasal secretory protein 1
A DNA sequence encoding the Mus musculus (Mouse) Vomeronasal secretory protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7718-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_034824.1 | |
Vomeronasal secretory protein 1 | |
Research Use Only | |
Lcn3 | |
Recombinant Protein | |
E. coli | |
QDSSFLAFNN GNFSGKWFLK ALVSEDDIPI NKVSPMLILV LNNGDIELSI THMIYDQCLE VTTILEKTDV PGQYLAFEGK THLQVQLSSV KGHYMLYCDG EIEGMRFLMT QLIGRDPQEN LEALEEFKVF TQIKGLVAEN LVILEQMEKC EPESFYELPS RPSE | |
Greater than 90% as determined by SDS-PAGE. |
16820 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |