missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Vomeronasal secretory protein 1
A DNA sequence encoding the Mus musculus (Mouse) Vomeronasal secretory protein 1, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_034824.1 |
---|---|
Gene ID (Entrez) | 16820 |
Name | Vomeronasal secretory protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16000391
|
enQuireBio™
QP7718-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
16020391
|
enQuireBio™
QP7718-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
16010391
|
enQuireBio™
QP7718-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16030391
|
enQuireBio™
QP7718-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16040391
|
enQuireBio™
QP7718-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16050391
|
enQuireBio™
QP7718-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_034824.1 | |
Vomeronasal secretory protein 1 | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
16820 | |
Research Use Only | |
Lcn3 | |
Recombinant Protein | |
E. coli | |
QDSSFLAFNN GNFSGKWFLK ALVSEDDIPI NKVSPMLILV LNNGDIELSI THMIYDQCLE VTTILEKTDV PGQYLAFEGK THLQVQLSSV KGHYMLYCDG EIEGMRFLMT QLIGRDPQEN LEALEEFKVF TQIKGLVAEN LVILEQMEKC EPESFYELPS RPSE | |
Greater than 90% as determined by SDS-PAGE. |