Learn More
enQuireBio™ Recombinant Mouse TECTB Protein
A DNA sequence encoding the Mus musculus (Mouse) TECTB, was expressed in the hosts and tags indicated.
469.00€ - 1705.00€
Specifications
Accession Number | NP_033374.2 |
---|---|
Gene ID (Entrez) | 21684 |
Name | TECTB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039141
|
enQuireBio™
QP9985-IV-100UG |
100 μg |
1705.00€
100µg |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
16059141
|
enQuireBio™
QP9985-IV-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16049141
|
enQuireBio™
QP9985-IV-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_033374.2 | |
TECTB Protein | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
21684 | |
Research Use Only | |
Tectb | |
Recombinant Protein | |
E. coli | |
KSCTPNKADV ILVFCYPKTI ITKIPECPYG WEVHQLALGG LCYNGVHEGG YYQFVIPDLS PKNKSYCGTQ SEYKPPIYHF YSHIVSNDST VIVKNQPVNY SFSCTYHSTY LVNQAAFDQR VATVHVKNGS MGTFESQLSL NFYTNAKFST KKEAPFVLET SEIGSDLFAG VEAKGLSVRF KVVLNSCWAT PSADFMYPLQ WQLINKGCPT DETVLVHENG KDHRATFQFN AFRFQNIPKL SKVWLHCETF ICDSEKLSCP VNCDKRKRML RDQTGGVLVV ELSLRSRA | |
Greater than 83.7% as determined by SDS-PAGE. |