missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus

Product Code. 18157800
Click to view available options
Quantity:
200 μg
25 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT .
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT with BSA as a carrier protein.
Unit Size:
200µg
25µg
This item is not returnable. View return policy

Product Code. 18157800

Brand: R&D Systems™ 464SH025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus is derived from E. coli. The Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT .
Gene ID (Entrez) 20423
Molecular Weight (g/mol) 19.8 kDa
Name Sonic Hedgehog/Shh (C25II), N-Terminus
Quantity 25 μg
Source E. coli-derived mouse Sonic Hedgehog/Shh protein Cys25-Gly198 (Cys25Ile-Ile),with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.