missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SAMHD1 Protein
A DNA sequence encoding the Mus musculus (Mouse) SAMHD1, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_001132992.1 |
---|---|
Gene ID (Entrez) | 56045 |
Name | SAMHD1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16004820
|
enQuireBio™
QP6650-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16024820
|
enQuireBio™
QP6650-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16014820
|
enQuireBio™
QP6650-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16034820
|
enQuireBio™
QP6650-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16044820
|
enQuireBio™
QP6650-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16054820
|
enQuireBio™
QP6650-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001132992.1 | |
SAMHD1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
56045 | |
Research Use Only | |
SAMHD1 | |
Recombinant Protein | |
E. coli | |
DIMITDAFLK ADPYVEITGT AGKKFRISTA IDDMEAFTKL TDNIFLEVLH STDPQLSEAQ SILRNIECRN LYKYLGETQP KREKIRKEEY ERLPQEVAKA KPEKAPDVEL KAEDFIVDVI NVDYGMEDKN PIDRVHFYCK SNSKQAVRIN KEQVSQLLPE KFAEQLIRVY CKKKDGKSLD AAGKHFVQWC ALRDFTKPQD GDIIAPLITP LKWNNKTSSC LQEVSKVKTC LK | |
Greater than 90% as determined by SDS-PAGE. |