missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SAMHD1 Protein

A DNA sequence encoding the Mus musculus (Mouse) SAMHD1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6650-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044820

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001132992.1
SAMHD1 Protein
Research Use Only
SAMHD1
Recombinant Protein
E. coli
DIMITDAFLK ADPYVEITGT AGKKFRISTA IDDMEAFTKL TDNIFLEVLH STDPQLSEAQ SILRNIECRN LYKYLGETQP KREKIRKEEY ERLPQEVAKA KPEKAPDVEL KAEDFIVDVI NVDYGMEDKN PIDRVHFYCK SNSKQAVRIN KEQVSQLLPE KFAEQLIRVY CKKKDGKSLD AAGKHFVQWC ALRDFTKPQD GDIIAPLITP LKWNNKTSSC LQEVSKVKTC LK
Greater than 90% as determined by SDS-PAGE.
56045
500 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers