missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Resistin Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Mouse Resistin is used at 0.5μg/mL, it binds to Biotinylated Recombinant Mouse Leptin with an ED50 of 50-300 ng/mL.
The Recombinant Mouse Resistin Protein is derived from E. coli. The Recombinant Mouse Resistin Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 57264 |
| Molecular Weight (g/mol) | 10 kDa (monomer) |
| Name | Resistin |
| Quantity | 50 μg |
| Source | E. coli-derived mouse Resistin protein Ser21-Ser114 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Monomer and disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido