missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Nus1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Nus1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9884-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16078911

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_084526.1
Nus1 Protein
Research Use Only
Nus1
Recombinant Protein
Yeast
SWLRVRFGTW NWIWRRCCRA ASAAVLAPLG FTLRKPRAVG RNRRHHRHPH GGPGPGPGPA ATHPRLRWRA DVRSLQKLPV HMGLLVTEEV QEPSFSD
Greater than 90% as determined by SDS-PAGE.
52014
50 μg
Not determined.
Cytoplasmic Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers