missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nus1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nus1, was expressed in the hosts and tags indicated.
245.00€ - 2645.00€
Specifications
Accession Number | NP_084526.1 |
---|---|
Gene ID (Entrez) | 52014 |
Name | Nus1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028911
|
enQuireBio™
QP9884-YE-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16068911
|
enQuireBio™
QP9884-YE-500UG |
500 μg |
2195.00€
500µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16038911
|
enQuireBio™
QP9884-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058911
|
enQuireBio™
QP9884-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16078911
|
enQuireBio™
QP9884-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048911
|
enQuireBio™
QP9884-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_084526.1 | |
Nus1 Protein | |
Not determined. | |
Cytoplasmic Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
52014 | |
Research Use Only | |
Nus1 | |
Recombinant Protein | |
Yeast | |
SWLRVRFGTW NWIWRRCCRA ASAAVLAPLG FTLRKPRAVG RNRRHHRHPH GGPGPGPGPA ATHPRLRWRA DVRSLQKLPV HMGLLVTEEV QEPSFSD | |
Greater than 90% as determined by SDS-PAGE. |