missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nus1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nus1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9884-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_084526.1 | |
Nus1 Protein | |
Research Use Only | |
Nus1 | |
Recombinant Protein | |
Yeast | |
SWLRVRFGTW NWIWRRCCRA ASAAVLAPLG FTLRKPRAVG RNRRHHRHPH GGPGPGPGPA ATHPRLRWRA DVRSLQKLPV HMGLLVTEEV QEPSFSD | |
Greater than 90% as determined by SDS-PAGE. |
52014 | |
500 μg | |
Not determined. | |
Cytoplasmic Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |