missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Neuregulin-1/NRG1 Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
- Derived from NS0
- Validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | XP_006509143 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 3084 |
| Molecular Weight (g/mol) | 42 to 49 kDa, reducing conditions |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse Neuregulin-1/NRG1 protein Ser20-Lys246, with a C-terminal 6-His tag |
| Storage Requirements | 12 months from date of receipt, -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution 3 months, -20 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | < 0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Species | Mouse |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction