missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Netrin-1 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS and EDTA with Trehalose.
Unit Size:
25µg
Description
The Recombinant Mouse Netrin-1 Protein is derived from NS0. The Recombinant Mouse Netrin-1 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and EDTA with Trehalose. |
| Gene ID (Entrez) | 18208 |
| Molecular Weight (g/mol) | 68.2 kDa |
| Name | Netrin-1 |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Netrin-1 protein, (N-terminus) Mouse Netrin-1 (Val22-Ala603) Accession &Num; AAC52971 IEGR GGGSGGGSGGGS 10-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction