missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse MMP-12 Protein, CF

Product Code. 18757098
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
This item is not returnable. View return policy

Product Code. 18757098

Brand: R&D Systems™ 3467MPB020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave a fluorogenic peptide substrate Mca-KPLGL-Dpa-AR-NH2 (Catalog # ES010 ). The specific activity is >100 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number EDL24933
Characterization Enzyme Activity
For Use With (Application) Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered solution in MES, NaCl, CaCl2, CHAPS, ZnSO4 and PEG.
Gene ID (Entrez) 4321
Molecular Weight (g/mol) M.W. (Observed): 50-62 kDa, reducing conditions; M.W. (theoretical): 52 kDa
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived mouse MMP-12 protein Ala18-Cys462
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME,mgC138506, MME, MMP12, MMP-12
Product Type Recombinant Protein
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 0.25 mg/mL in 10 mM MES, 0.1 M NaCl, 100 μM CaCl2, 0.1% CHAPS, 1 μM ZnSO4 and 0.1% PEG, pH 6.0
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein MMP-12
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.