Learn More
enQuireBio™ Recombinant Mouse Major urinary protein 11 Protein
A DNA sequence encoding the Mus musculus (Mouse) Major urinary protein 11, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_001157998.1 |
---|---|
Gene ID (Entrez) | 100039028 |
Name | Major urinary protein 11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008440
|
enQuireBio™
QP7360-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
16028440
|
enQuireBio™
QP7360-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
16018440
|
enQuireBio™
QP7360-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16038440
|
enQuireBio™
QP7360-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048440
|
enQuireBio™
QP7360-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058440
|
enQuireBio™
QP7360-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001157998.1 | |
Major urinary protein 11 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
100039028 | |
Research Use Only | |
Mup11 | |
Recombinant Protein | |
E. coli | |
REKINGEWHT IILASDKREK IEDNGNFRLF LEQIHVLENS LVLKFHTVRD EECSELSMVA DKTEKAGEYS VTYDGFNTFT IPKTDYDNFL MAHLINEKDG ETFQLMGLYG REPDLSSDIK ERFAQLCEEH GILRENIIDL SNANRCLQAR E | |
Greater than 81.5% as determined by SDS-PAGE. |