missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Lefty-1 Protein
Description
The Recombinant Mouse Lefty-1 Protein is derived from NS0. The Recombinant Mouse Lefty-1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 10637 |
| Molecular Weight (g/mol) | 26.9 kDa |
| Name | Lefty-1 |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Lefty-1 protein Mouse Lefty-1 Proregion (Leu22-Arg135) Accession &Num; NP_034224 6-His tag Mouse Lefty mature (Leu136-Pro368) Accession &Num; NP_034224 6-His tag Mouse Lefty mature (Leu136-Pro368) Accession &Num; NP_034224 N-terminus C-terminus A short DNA sequence encoding 6 histidine residues was inserted into the DNA sequence encoding mouse Lefty-1 between Arg135 and Leu136. Two peptides corresponding to mouse Lefty-1 propeptide and mature mouse Lefty-1 are present in the recombinant preparation. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?