missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Interferon epsilon Protein

A DNA sequence encoding the Mus musculus (Mouse) Interferon epsilon, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7798-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16060861

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_796322.1
Interferon epsilon Protein
Research Use Only
Ifne
Recombinant Protein
E. coli
LEPKRIPFQL WMNRESLQLL KPLPSSSVQQ CLAHRKNFLL PQQPVSPHQY QEGQVLAVVH EILQQIFTLL QTHGTMGIWE ENHIEKVLAA LHRQLEYVES LGGLNAAQKS GGSSAQNLRL QIKAYFRRIH DYLENQRYSS CAWIIVQTEI HRCMFFVFRF TTWLSRQDPD P
Greater than 90% as determined by SDS-PAGE.
230405
200 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers