missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Interferon epsilon Protein
A DNA sequence encoding the Mus musculus (Mouse) Interferon epsilon, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_796322.1 |
---|---|
Gene ID (Entrez) | 230405 |
Name | Interferon epsilon Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16030861
|
enQuireBio™
QP7798-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16050861
|
enQuireBio™
QP7798-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16040861
|
enQuireBio™
QP7798-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16060861
|
enQuireBio™
QP7798-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16070861
|
enQuireBio™
QP7798-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16080861
|
enQuireBio™
QP7798-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_796322.1 | |
Interferon epsilon Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
230405 | |
Research Use Only | |
Ifne | |
Recombinant Protein | |
E. coli | |
LEPKRIPFQL WMNRESLQLL KPLPSSSVQQ CLAHRKNFLL PQQPVSPHQY QEGQVLAVVH EILQQIFTLL QTHGTMGIWE ENHIEKVLAA LHRQLEYVES LGGLNAAQKS GGSSAQNLRL QIKAYFRRIH DYLENQRYSS CAWIIVQTEI HRCMFFVFRF TTWLSRQDPD P | |
Greater than 90% as determined by SDS-PAGE. |