missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Insulin-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Insulin-1, was expressed in the hosts and tags indicated.
287.00€ - 3175.00€
Specifications
Accession Number | NP_032412.3 |
---|---|
Gene ID (Entrez) | 16333 |
Name | Insulin-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028531
|
enQuireBio™
QP9628-YE-100UG |
100 μg |
920.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16068531
|
enQuireBio™
QP9628-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16038531
|
enQuireBio™
QP9628-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058531
|
enQuireBio™
QP9628-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16078531
|
enQuireBio™
QP9628-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048531
|
enQuireBio™
QP9628-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_032412.3 | |
Insulin-1 Protein | |
Not determined. | |
Full. Length. of. Mature. Protein. | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
16333 | |
Research Use Only | |
Ins1 | |
Recombinant Protein | |
Yeast | |
FVKQHLCGPH LVEALYLVCG ERGFFYTPKS RREVEDPQVE QLELGGSPGD LQTLALEVAR QKRGIVDQCC TSICSLYQLE NYCN | |
Greater than 80% as determined by SDS-PAGE. |