missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Insulin-1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Insulin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9628-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048531

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_032412.3
Insulin-1 Protein
Research Use Only
Ins1
Recombinant Protein
Yeast
FVKQHLCGPH LVEALYLVCG ERGFFYTPKS RREVEDPQVE QLELGGSPGD LQTLALEVAR QKRGIVDQCC TSICSLYQLE NYCN
Greater than 80% as determined by SDS-PAGE.
16333
1 mg
Not determined.
Full. Length. of. Mature. Protein.
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers