missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL7R / IL7RA Protein
A DNA sequence encoding the Mus musculus (Mouse) IL7R / IL7RA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6226-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032398.3 | |
IL7R / IL7RA Protein | |
Research Use Only | |
Il7r | |
Recombinant Protein | |
E. coli | |
ESGNAQDGDL EDADADDHSF WCHSQLEVDG SQHLLTCAFN DSDINTANLE FQICGALLRV KCLTLNKLQD IYFIKTSEFL LIGSSNICVK LGQKNLTCKN MAINTIVKAE APSDLKVVYR KEANDFLVTF NAPHLKKKYL KKVKHDVAYR PARGESNWTH VSLFHTRTTI PQRKLRPKAM YEIKVRSIPH NDYFKGFWSE WSPSSTFETP EPKNQGGWD | |
Greater than 90% as determined by SDS-PAGE. |
16197 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |