missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL7R / IL7RA Protein
A DNA sequence encoding the Mus musculus (Mouse) IL7R / IL7RA, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_032398.3 |
---|---|
Gene ID (Entrez) | 16197 |
Name | IL7R / IL7RA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16001740
|
enQuireBio™
QP6226-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16021740
|
enQuireBio™
QP6226-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16011740
|
enQuireBio™
QP6226-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16031740
|
enQuireBio™
QP6226-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16041740
|
enQuireBio™
QP6226-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16051740
|
enQuireBio™
QP6226-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_032398.3 | |
IL7R / IL7RA Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
16197 | |
Research Use Only | |
Il7r | |
Recombinant Protein | |
E. coli | |
ESGNAQDGDL EDADADDHSF WCHSQLEVDG SQHLLTCAFN DSDINTANLE FQICGALLRV KCLTLNKLQD IYFIKTSEFL LIGSSNICVK LGQKNLTCKN MAINTIVKAE APSDLKVVYR KEANDFLVTF NAPHLKKKYL KKVKHDVAYR PARGESNWTH VSLFHTRTTI PQRKLRPKAM YEIKVRSIPH NDYFKGFWSE WSPSSTFETP EPKNQGGWD | |
Greater than 90% as determined by SDS-PAGE. |