Learn More
enQuireBio™ Recombinant Mouse IFNAR2 / IFNABR Protein
A DNA sequence encoding the Mus musculus (Mouse) IFNAR2 / IFNABR, was expressed in the hosts and tags indicated.
245.00€ - 2645.00€
Specifications
Accession Number | NP_001103968.1 |
---|---|
Gene ID (Entrez) | 15976 |
Name | IFNAR2 / IFNABR Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057831
|
enQuireBio™
QP9199-YE-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16097831
|
enQuireBio™
QP9199-YE-500UG |
500 μg |
2195.00€
500µg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16067831
|
enQuireBio™
QP9199-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16087831
|
enQuireBio™
QP9199-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16007841
|
enQuireBio™
QP9199-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16077831
|
enQuireBio™
QP9199-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001103968.1 | |
IFNAR2 / IFNABR Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
15976 | |
Research Use Only | |
Ifnar2 | |
Recombinant Protein | |
Yeast | |
SLETITPSAF DGYPDEPCTI NITIRNSRLI LSWELENKSG PPANYTLWYT VMSKDENLTK VKNCSDTTKS SCDVTDKWLE GMESYVVAIV IVHRGDLTVC RCSDYIVPAN APLEPPEFEI VGFTDHINVT MEFPPVTSKI IQEKMKTTPF VIKEQIGDSV RKKHEPKVNN VTGNFTFVLR DLLPKTNYCV SLYFDDDPAI KSPLKCIVLQ PGQESGLSES A | |
Greater than 90% as determined by SDS-PAGE. |