missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IFN-alpha 7/IFNA7 Protein

Product Code. 18776616
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18776616

Brand: R&D Systems™ 10208IJ010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured in an anti-viral assay using L-929 mouse fibroblast cells infected with encephalomyocarditis (EMC) virus.

  • The ED50 for this effect is <1.5 ng/mL.
  • Storage:
    • Use a manual defrost freezer and avoid repeated freeze-thaw cycles
    • 12 months from date of receipt, -20 to -70°C as supplied
    • 1 month, 2 to 8°C under sterile conditions after reconstitution
    • 3 months, -20 to -70°C under sterile conditions after reconstitution
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P06799
For Use With (Application) Bioactivity
Gene ID (Entrez) 3444
Molecular Weight (g/mol) 18-24 kDa, under reducing conditions
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse IFN-alpha 7/IFNA7 protein Cys24-Glu190
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles
Endotoxin Concentration < 0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 100 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Purity or Quality Grade > 95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.