missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Histone H3-like centromeric protein 1

A DNA sequence encoding the Mus musculus (Mouse) Histone H3-like centromeric protein A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9060-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027551

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001289058.1
Histone H3-like centromeric protein 1
Research Use Only
Cenpa
Recombinant Protein
Yeast
MGPRRKPQTP RRRPSSPAPG PSRQSSSVGS QTLRRRQKFM WLKEIKTLQK STDLLFRKKP FSMVVREICE KFSRGVDFWW QAQALLALQE AAEAFLIHLF EDAYLLSLHA GRVTLFPKDI QLTRRIRGFE GGLP
Greater than 90% as determined by SDS-PAGE.
12615
10 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers