missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Histone H3-like centromeric protein 1
A DNA sequence encoding the Mus musculus (Mouse) Histone H3-like centromeric protein A, was expressed in the hosts and tags indicated.
245.00€ - 2645.00€
Specifications
Accession Number | NP_001289058.1 |
---|---|
Gene ID (Entrez) | 12615 |
Name | Histone H3-like centromeric protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017551
|
enQuireBio™
QP9060-YE-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16057551
|
enQuireBio™
QP9060-YE-500UG |
500 μg |
2195.00€
500µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16027551
|
enQuireBio™
QP9060-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16047551
|
enQuireBio™
QP9060-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16067551
|
enQuireBio™
QP9060-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16037551
|
enQuireBio™
QP9060-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001289058.1 | |
Histone H3-like centromeric protein 1 | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
12615 | |
Research Use Only | |
Cenpa | |
Recombinant Protein | |
Yeast | |
MGPRRKPQTP RRRPSSPAPG PSRQSSSVGS QTLRRRQKFM WLKEIKTLQK STDLLFRKKP FSMVVREICE KFSRGVDFWW QAQALLALQE AAEAFLIHLF EDAYLLSLHA GRVTLFPKDI QLTRRIRGFE GGLP | |
Greater than 90% as determined by SDS-PAGE. |