missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse HAPLN1 Protein
A DNA sequence encoding the Mus musculus (Mouse) HAPLN1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9171-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_038528.3 | |
HAPLN1 Protein | |
Research Use Only | |
Hapln1 | |
Recombinant Protein | |
Yeast | |
ISVCWADHHL SDSYTPPDQD RVIHIQAENG PRLLVEAEQA KVFSHRGGNV TLPCKFYRDP TAFGSGIHKI RIKWTKLTSD YLREVDVFVS MGYHKKTYGG YQGRVFLKGG SDNDASLVIT DLTLEDYGRY KCEVIEGLED DTAVVALELQ GVVFPYFPRL GRYNLNFHEA RQACLDQDAV IASFDQLYDA WRGGLDWCNA GWLSDGSVQY PITKPREPCG GQNTVPGVRN YGFWDKDKSR YDVFCFTSNF NGRFYYLIHP TKLTYDEAVQ ACLNDGAQIA KVGQIFAAWK LLGYDRCDAG WLADGSVRYP ISRPRRRCSP TEAAVRFVGF PDKKHKLYGV YCFRAYN | |
Greater than 90% as determined by SDS-PAGE. |
12950 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |