missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse HAPLN1 Protein
A DNA sequence encoding the Mus musculus (Mouse) HAPLN1, was expressed in the hosts and tags indicated.
287.00€ - 3175.00€
Specifications
Accession Number | NP_038528.3 |
---|---|
Gene ID (Entrez) | 12950 |
Name | HAPLN1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037761
|
enQuireBio™
QP9171-YE-100UG |
100 μg |
920.00€
100µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16077761
|
enQuireBio™
QP9171-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16047761
|
enQuireBio™
QP9171-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16067761
|
enQuireBio™
QP9171-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16087761
|
enQuireBio™
QP9171-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16057761
|
enQuireBio™
QP9171-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_038528.3 | |
HAPLN1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
12950 | |
Research Use Only | |
Hapln1 | |
Recombinant Protein | |
Yeast | |
ISVCWADHHL SDSYTPPDQD RVIHIQAENG PRLLVEAEQA KVFSHRGGNV TLPCKFYRDP TAFGSGIHKI RIKWTKLTSD YLREVDVFVS MGYHKKTYGG YQGRVFLKGG SDNDASLVIT DLTLEDYGRY KCEVIEGLED DTAVVALELQ GVVFPYFPRL GRYNLNFHEA RQACLDQDAV IASFDQLYDA WRGGLDWCNA GWLSDGSVQY PITKPREPCG GQNTVPGVRN YGFWDKDKSR YDVFCFTSNF NGRFYYLIHP TKLTYDEAVQ ACLNDGAQIA KVGQIFAAWK LLGYDRCDAG WLADGSVRYP ISRPRRRCSP TEAAVRFVGF PDKKHKLYGV YCFRAYN | |
Greater than 90% as determined by SDS-PAGE. |