missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GRK2 / ADRBK1 Protein
A DNA sequence encoding the Mus musculus (Mouse) GRK2 / ADRBK1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8014-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001277747.1 | |
GRK2 / ADRBK1 Protein | |
Research Use Only | |
Adrbk1 | |
Recombinant Protein | |
E. coli | |
MADLEAVLAD VSYLMAMEKS KATPAARASK KILLPEPSIR SVMQKYLEDR GEVTFEKIFS QKLGYLLFRD FCLNHLEEAK PLVEFYEEIK KYEKLETEEE RVVRSREIFD SYIMKELLAC SHPFSKNATE HVQGHLVKKQ VPPDLFQPYI EEICQNLRGD VFQKFIESDK FTRFCQWKNV ELNIHLTMND FSVHRIIGRG GFGEVYGCRK ADTGKMYAMK CLDKKRIKMK QGETLALNER IMLSLVSTGD CPFIVCMSYA FHTPDKLSFI LDLMNGGDLH YHLSQHGVFS EADMRFYAAE IILGLEHMHN RFVVYRDLKP ANILLDEHGH VRISDLGLAC DFSKKRPHAS VGTHGYMAPE VLQKGVAYDS SADWFSLGCM LFKLLRGHSP FRQHKTKDKH EIDRMTLTMA VELPDSFSPE LRSLLEGLLQ RDVNRRLGCL GRGAQEVKES PFFRSLDWQM VFLQKYPPPL IPPRGEVNAA DAFDIGSFDE EDTKGIKLLD SDQELYRNFP LTISERWQQE VAETVFDTIN AETDRLEARK KAKNKQLGHE EDYALGKDCI VHGYMSKMGN PFLTQWQRRY FYLFPNRLEW RGEGEAPQSL LTMEEIQSVE ETQIKERKCL LLKIRGGKQF VLQCDSDPEL VQWKKELRDA YREAQQLVQR VPKMKNKPRS PVVELSKVPL IQRGSANGL | |
Greater than 90% as determined by SDS-PAGE. |
110355 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |