missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GRK2 / ADRBK1 Protein
A DNA sequence encoding the Mus musculus (Mouse) GRK2 / ADRBK1, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_001277747.1 |
---|---|
Gene ID (Entrez) | 110355 |
Name | GRK2 / ADRBK1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022161
|
enQuireBio™
QP8014-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16042161
|
enQuireBio™
QP8014-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16032161
|
enQuireBio™
QP8014-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16052161
|
enQuireBio™
QP8014-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16062161
|
enQuireBio™
QP8014-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16072161
|
enQuireBio™
QP8014-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001277747.1 | |
GRK2 / ADRBK1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
110355 | |
Research Use Only | |
Adrbk1 | |
Recombinant Protein | |
E. coli | |
MADLEAVLAD VSYLMAMEKS KATPAARASK KILLPEPSIR SVMQKYLEDR GEVTFEKIFS QKLGYLLFRD FCLNHLEEAK PLVEFYEEIK KYEKLETEEE RVVRSREIFD SYIMKELLAC SHPFSKNATE HVQGHLVKKQ VPPDLFQPYI EEICQNLRGD VFQKFIESDK FTRFCQWKNV ELNIHLTMND FSVHRIIGRG GFGEVYGCRK ADTGKMYAMK CLDKKRIKMK QGETLALNER IMLSLVSTGD CPFIVCMSYA FHTPDKLSFI LDLMNGGDLH YHLSQHGVFS EADMRFYAAE IILGLEHMHN RFVVYRDLKP ANILLDEHGH VRISDLGLAC DFSKKRPHAS VGTHGYMAPE VLQKGVAYDS SADWFSLGCM LFKLLRGHSP FRQHKTKDKH EIDRMTLTMA VELPDSFSPE LRSLLEGLLQ RDVNRRLGCL GRGAQEVKES PFFRSLDWQM VFLQKYPPPL IPPRGEVNAA DAFDIGSFDE EDTKGIKLLD SDQELYRNFP LTISERWQQE VAETVFDTIN AETDRLEARK KAKNKQLGHE EDYALGKDCI VHGYMSKMGN PFLTQWQRRY FYLFPNRLEW RGEGEAPQSL LTMEEIQSVE ETQIKERKCL LLKIRGGKQF VLQCDSDPEL VQWKKELRDA YREAQQLVQR VPKMKNKPRS PVVELSKVPL IQRGSANGL | |
Greater than 90% as determined by SDS-PAGE. |