missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Glutaminyl cyclase / QPCT Protein

Product Code. p-7153355
Click to view available options
Quantity:
100 μg
20 μg
50 μg
Unit Size:
100µg
20µg
50µg
This item is not returnable. View return policy

Product Code. 16004230

Brand: enQuireBio™ QP6567ma20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Glutaminyl cyclase / QPCT, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001303658.1
Gene ID (Entrez) 70536
Name Glutaminyl cyclase / QPCT Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol QPCT
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Mammal
Protein Tag His
Sequence AWTQEKNHHQ PAHLNSSSLQ QVAEGTSISE MWQNDLRPLL IERYPGSPGS YSARQHIMQR IQRLQAEWVV EVDTFLSRTP YGYRSFSNII STLNPEAKRH LVLACHYDSK YFPRWDSRVF VGATDSAVPC AMMLELARAL DKKLHSLKDV SGSKPDLSLR LIFFDGEEAF HHWSPQDSLY GSRHLAQKMA SSPHPPGSRG TNQLDGMDLL VLLDLIGAAN PTFPNFFPKT TRWFNRLQAI EKELYELGLL KDHSLERKYF QNFGYGNIIQ DDHIPFLRKG VPVLHLIASP FPEVWHTMDD NEENLHASTI DNLNKIIQVF VLEYLHL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.