missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Ephrin-B2 Fc Chimera Protein
Click to view available options
:
200ug; Carrier Free
Description
The Recombinant Mouse Ephrin-B2 Fc Chimera Protein is derived from NS0. The Recombinant Mouse Ephrin-B2 Fc Chimera Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 13642 |
| Molecular Weight (g/mol) | 49.6 kDa (monomer) |
| Name | Ephrin-B2 Fc Chimera |
| Quantity | 200μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Ephrin-B2 protein, (N-terminus) Mouse Ephrin-B2 (Arg27-Ala227) Accession # AAA82934 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto