missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CD8 alpha Protein

Product Code. 18178150
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18178150

Brand: R&D Systems™ 9135CD050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse CD8 alpha Protein is derived from NS0. The Recombinant Mouse CD8 alpha Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01731
For Use With (Application) Bioactivity
Gene ID (Entrez) 925
Molecular Weight (g/mol) Observed M.W.: 32-41kDa, reducing conditions, Theoretical M.W.: 20kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse CD8 protein Lys28-Tyr196,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name CD8
Gene Symbol Cd8a
Biological Activity Measured by its ability of the immobilized protein to support the adhesion of Jurkat human acute T cell leukemia cells. The ED50 for this effect is typically 1-5μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Lys28-Tyr196, with a C-terminal 10-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.