missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CCL12 / MCP-5 Protein

A DNA sequence encoding the Mus musculus (Mouse) CCL12 / MCP-5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8648-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16075751

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_035461.2
CCL12 / MCP-5 Protein
Research Use Only
Ccl12
Recombinant Protein
E. coli
GPDAVSTPVT CCYNVVKQKI HVRKLKSYRR ITSSQCPREA VIFRTILDKE ICADPKEKWV KNSINHLDKT SQTFILEPSC LG
Greater than 90% as determined by SDS-PAGE.
20293
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers